Product Details

SNP ID
rs2437008
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:47028933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAAGCAGCTTCCTCCCGGAACT[C/T]TGGTGAAGAGAGTGCTTCTGGCTTT
Phenotype
MIM: 614697 MIM: 609522
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KATNAL2 PubMed Links

Gene Details

Gene
KATNAL2
Gene Name
katanin catalytic subunit A1 like 2
There are no transcripts associated with this gene.

Gene
TCEB3B
Gene Name
transcription elongation factor B subunit 3B
There are no transcripts associated with this gene.

Gene
TCEB3C
Gene Name
transcription elongation factor B subunit 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145653.3 1146 Missense Mutation ACG,GCG T304A NP_663628.2
Gene
TCEB3CL
Gene Name
transcription elongation factor B subunit 3C like
There are no transcripts associated with this gene.

View Full Product Details