Product Details

SNP ID
rs2511225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66480960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATGAGAATGGGACAGTTTCAAGC[A/C]AAATCATTTATTGCTGTGGATACTT
Phenotype
MIM: 606818 MIM: 609827
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DPP3 PubMed Links

Gene Details

Gene
DPP3
Gene Name
dipeptidyl peptidase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256670.1 Intron NP_001243599.1
NM_005700.4 Intron NP_005691.2
NM_130443.3 Intron NP_569710.2
Gene
LOC101928069
Gene Name
uncharacterized LOC101928069
There are no transcripts associated with this gene.

Gene
PELI3
Gene Name
pellino E3 ubiquitin protein ligase family member 3
There are no transcripts associated with this gene.

View Full Product Details