Product Details
- SNP ID
-
rs2513829
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:98076685 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATATTCATATAGGATTGTTCAATC[C/T]AGTTCAGTCTCTCCAAGTAGCTCCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ERICH5
PubMed Links
Gene Details
- Gene
- ERICH5
- Gene Name
- glutamate rich 5
View Full Product Details