Product Details

SNP ID
rs2515637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47774336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCGTGCCACAGCCACATAGTCT[C/T]TGAAGGCAGGGGCCTTCTCATGACC
Phenotype
MIM: 600899
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRKDC PubMed Links

Gene Details

Gene
PRKDC
Gene Name
protein kinase, DNA-activated, catalytic polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081640.1 12188 Missense Mutation AAA,AGA K4044R NP_001075109.1
NM_006904.6 12188 Missense Mutation AAA,AGA K4075R NP_008835.5

View Full Product Details