Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001110798.1 | Intron | NP_001104268.1 | |||
NM_018527.3 | Intron | NP_060997.2 | |||
NM_024561.4 | Intron | NP_078837.3 | |||
XM_005266523.2 | Intron | XP_005266580.1 | |||
XM_006719866.2 | Intron | XP_006719929.1 | |||
XM_011535227.2 | Intron | XP_011533529.1 | |||
XM_011535228.1 | Intron | XP_011533530.1 | |||
XM_017020744.1 | Intron | XP_016876233.1 | |||
XM_017020745.1 | Intron | XP_016876234.1 | |||
XM_017020746.1 | Intron | XP_016876235.1 |