Product Details
- SNP ID
-
rs2540972
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:37205084 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGGAAGACAAAATCTGGAAGATAAC[A/G]TGTTATAATTTAAATATGTATCAGT
- Phenotype
-
MIM: 612828
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CEBPZ
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75355833] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEBPZ
- Gene Name
- CCAAT/enhancer binding protein zeta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005760.2 |
|
Intron |
|
|
NP_005751.2 |
- Gene
- CEBPZOS
- Gene Name
- CEBPZ opposite strand
View Full Product Details