Product Details
- SNP ID
-
rs2502729
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128211204 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAGTTTCCTCCCTTGCCCCCTAA[C/T]CCATTCCCCGCCCGACAGCATGAAA
- Phenotype
-
MIM: 611420
MIM: 602377
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CIZ1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113574484] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CIZ1
- Gene Name
- CDKN1A interacting zinc finger protein 1
There are no transcripts associated with this gene.
- Gene
- DNM1
- Gene Name
- dynamin 1
View Full Product Details