Product Details

SNP ID
rs2598414
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76071018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGCAAACTGAAAAAGGAGGGCA[C/T]TACTGGCCACTGGAAACTGCCAAAA
Phenotype
MIM: 608163 MIM: 603691 MIM: 604858 MIM: 610935
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC7 PubMed Links

Gene Details

Gene
EXOC7
Gene Name
exocyst complex component 7
There are no transcripts associated with this gene.

Gene
GALR2
Gene Name
galanin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003857.3 197 Intron NP_003848.1
XM_011525427.2 197 UTR 5 XP_011523729.1
Gene
SRP68
Gene Name
signal recognition particle 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001260502.1 197 Intron NP_001247431.1
NM_001260503.1 197 Intron NP_001247432.1
NM_014230.3 197 Intron NP_055045.2
Gene
ZACN
Gene Name
zinc activated ion channel
There are no transcripts associated with this gene.

View Full Product Details