Product Details

SNP ID
rs2555364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48127189 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTCAGTTCATAGTGCTGTGTGA[C/G]AAGTGCACGAAGAAACACAAGTGAA
Phenotype
MIM: 609802
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYEF2 PubMed Links

Gene Details

Gene
MYEF2
Gene Name
myelin expression factor 2
There are no transcripts associated with this gene.

Gene
SLC24A5
Gene Name
solute carrier family 24 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_205850.2 1294 Intron NP_995322.1
XM_017022079.1 1294 UTR 5 XP_016877568.1
XM_017022080.1 1294 UTR 5 XP_016877569.1

View Full Product Details