Product Details

SNP ID
rs2633381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130210586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTCTGGACAATGGTAATCCAAAC[A/G]CCTTATTTTGCTTATGTGGAAACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSMEM1 PubMed Links

Gene Details

Gene
SSMEM1
Gene Name
serine rich single-pass membrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145268.3 Intron NP_660311.1
XM_005250141.3 Intron XP_005250198.1
XM_011515794.2 Intron XP_011514096.1
XM_011515795.2 Intron XP_011514097.1
Gene
TMEM209
Gene Name
transmembrane protein 209
There are no transcripts associated with this gene.

View Full Product Details