Product Details
- SNP ID
-
rs2572427
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:11289480 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTTGTATACAAGTTTTACAGTTA[C/T]ACCTGGTTTTTTTTTTAATGGACCT
- Phenotype
-
MIM: 606260
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MTMR9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79799107] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MTMR9
- Gene Name
- myotubularin related protein 9
View Full Product Details