Product Details

SNP ID
rs2634164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10174435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATGAGATTAACATATTAATCAG[A/C]AGAGGAAAGTAGATTTCGCTGCATA
Phenotype
MIM: 602601
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OLR1 PubMed Links

Gene Details

Gene
OLR1
Gene Name
oxidized low density lipoprotein receptor 1
There are no transcripts associated with this gene.

Gene
TMEM52B
Gene Name
transmembrane protein 52B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079815.1 Intron NP_001073283.1
NM_153022.3 Intron NP_694567.1
XM_006719029.2 Intron XP_006719092.1

View Full Product Details