Product Details

SNP ID
rs2257796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142727249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCGCTCCGGGCGCGCTGCGCTCG[C/T]GAGTTCCCAGGAGGCCTGGCGGGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
THEM6
Gene Name
thioesterase superfamily member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016647.2 47 UTR 5 NP_057731.1
XM_005250955.2 47 UTR 5 XP_005251012.1

View Full Product Details