Product Details

SNP ID
rs2645643
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:110150641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCTCCGCCGTGGGACCTGGGT[C/T]CCCGCGCCGCTCCCCTGAGCCCCGA
Phenotype
MIM: 610299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A17 PubMed Links

Gene Details

Gene
SLC6A17
Gene Name
solute carrier family 6 member 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010898.2 132 UTR 5 NP_001010898.1
XM_006710643.2 132 Intron XP_006710706.1

View Full Product Details