Product Details

SNP ID
rs2615951
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6203074 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAATGTATGGGCTTCTAACTGAG[A/G]AGTAATCAGTCAGAAGTTTTCAGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C11orf42 PubMed Links

Gene Details

Gene
C11orf42
Gene Name
chromosome 11 open reading frame 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173525.2 78 Intron NP_775796.2
XM_011519926.2 78 UTR 5 XP_011518228.1
XM_011519927.2 78 UTR 5 XP_011518229.1
Gene
FAM160A2
Gene Name
family with sequence similarity 160 member A2
There are no transcripts associated with this gene.

Gene
OR52W1
Gene Name
olfactory receptor family 52 subfamily W member 1
There are no transcripts associated with this gene.

View Full Product Details