Product Details
- SNP ID
-
rs2648841
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:128150187 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTCTGATGAGTCACCACTGTGGT[G/T]GGGGGGAGTGGACAGTCAGCCAGAC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR1208
PubMed Links
Gene Details
- Gene
- MIR1208
- Gene Name
- microRNA 1208
There are no transcripts associated with this gene.
View Full Product Details