Product Details

SNP ID
rs2672594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:122468686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGTCCCAGTTGTTGCCTATATCA[A/T]GTGACCAGTGTGTGGCTTCTCTTGA
Phenotype
MIM: 602194
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HTRA1 PubMed Links

Gene Details

Gene
HTRA1
Gene Name
HtrA serine peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002775.4 Intron NP_002766.1

View Full Product Details