Product Details

SNP ID
rs2672785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1225711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGCCCTGTGGAGCCGAGCTGGG[A/G]GAATGCAGGGCACACCATGGATGGC
Phenotype
MIM: 600770
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUC5B PubMed Links

Gene Details

Gene
MUC5B
Gene Name
mucin 5B, oligomeric mucus/gel-forming
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002458.2 160 Missense Mutation GAG,GGG E34G NP_002449.2

View Full Product Details