Product Details

SNP ID
rs2723147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:23395338 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGCTGCTATTGATTTGTCTCTCC[A/G]TCACAGTAGAGAGGGGTAGAAAATG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHL29 PubMed Links

Gene Details

Gene
KLHL29
Gene Name
kelch like family member 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052920.1 Intron NP_443152.1
XM_006711929.3 Intron XP_006711992.1
XM_011532501.2 Intron XP_011530803.1
XM_017003264.1 Intron XP_016858753.1

View Full Product Details