Product Details

SNP ID
rs2696292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50172440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAGCGGGCCTGCACGGTGCGTG[A/C]ACATATGCATGTGTGTGTGCACATG
Phenotype
MIM: 608101 MIM: 600119
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HILS1 PubMed Links

Gene Details

Gene
HILS1
Gene Name
histone linker H1 domain, spermatid-specific 1 (pseudogene)
There are no transcripts associated with this gene.

Gene
SGCA
Gene Name
sarcoglycan alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000023.3 Intron NP_000014.1
NM_001135697.2 Intron NP_001129169.1
XM_011525120.2 Intron XP_011523422.2
XM_011525121.2 Intron XP_011523423.2
XM_011525122.2 Intron XP_011523424.2
XM_011525123.2 Intron XP_011523425.2
XM_011525124.2 Intron XP_011523426.1
XM_017024950.1 Intron XP_016880439.1

View Full Product Details