Product Details

SNP ID
rs2723650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6453647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTAAGGGAATGAGGGAGTTTTC[A/G]CAGAAGAGCTGATAAAACTGGAAGC
Phenotype
MIM: 605493
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM3 PubMed Links

Gene Details

Gene
TRIM3
Gene Name
tripartite motif containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001248006.1 Intron NP_001234935.1
NM_001248007.1 Intron NP_001234936.1
NM_006458.3 Intron NP_006449.2
NM_033278.3 Intron NP_150594.2
XM_011519844.1 Intron XP_011518146.1
XM_011519845.2 Intron XP_011518147.1
XM_017017098.1 Intron XP_016872587.1
XM_017017099.1 Intron XP_016872588.1

View Full Product Details