Product Details
- SNP ID
-
rs2723852
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:104118773 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCCACTCACCCATGCAAAAGGT[C/G]TGTACACGCAATGATGTCTGATGTT
- Phenotype
-
MIM: 607926
MIM: 189904
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HCFC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78519510] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCFC2
- Gene Name
- host cell factor C2
There are no transcripts associated with this gene.
- Gene
- NFYB
- Gene Name
- nuclear transcription factor Y subunit beta
View Full Product Details