Product Details

SNP ID
rs2697063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:116216148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAAGAGAGTGATAAATTGGTA[A/C]CATGGAAGCTACTTCCTGTGTTCTT
Phenotype
MIM: 606085
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TES PubMed Links

Gene Details

Gene
TES
Gene Name
testin LIM domain protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015641.3 Intron NP_056456.1
NM_152829.2 Intron NP_690042.1
XM_005250258.1 Intron XP_005250315.1

View Full Product Details