Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039083.3 | Intron | NP_001034172.3 | |||
NM_001103154.1 | Intron | NP_001096624.1 | |||
XM_005256918.4 | Intron | XP_005256975.1 | |||
XM_011524160.2 | Intron | XP_011522462.1 | |||
XM_011524161.2 | Intron | XP_011522463.1 | |||
XM_011524162.2 | Intron | XP_011522464.1 | |||
XM_011524163.2 | Intron | XP_011522465.1 | |||
XM_011524164.2 | Intron | XP_011522466.1 | |||
XM_011524165.2 | Intron | XP_011522467.1 | |||
XM_011524166.2 | Intron | XP_011522468.1 | |||
XM_011524167.2 | Intron | XP_011522469.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014834.4 | Intron | NP_055649.4 | |||
XM_005257879.4 | Intron | XP_005257936.1 | |||
XM_005257880.4 | Intron | XP_005257937.1 | |||
XM_005257882.4 | Intron | XP_005257939.1 | |||
XM_005257883.4 | Intron | XP_005257940.1 | |||
XM_005257884.4 | Intron | XP_005257941.1 | |||
XM_006722210.3 | Intron | XP_006722273.1 | |||
XM_006722211.3 | Intron | XP_006722274.1 | |||
XM_011525539.2 | Intron | XP_011523841.1 | |||
XM_017025472.1 | Intron | XP_016880961.1 | |||
XM_017025473.1 | Intron | XP_016880962.1 |