Product Details

SNP ID
rs2214472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14733893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAGTAGAGGTACCAGGTTGCCAA[C/T]AATGGTTACTTGGGGGGTAGCATTC
Phenotype
MIM: 606100 MIM: 611811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADGRE2 PubMed Links

Gene Details

Gene
ADGRE2
Gene Name
adhesion G protein-coupled receptor E2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271052.1 5271 UTR 3 NP_001257981.1
NM_013447.3 5271 UTR 3 NP_038475.2
XM_011527948.2 5271 Intron XP_011526250.1
XM_011527949.2 5271 Intron XP_011526251.1
XM_011527951.2 5271 Intron XP_011526253.1
XM_011527952.2 5271 Intron XP_011526254.1
XM_011527953.2 5271 Intron XP_011526255.1
XM_011527954.2 5271 Intron XP_011526256.1
XM_011527955.2 5271 Intron XP_011526257.1
XM_017026726.1 5271 Intron XP_016882215.1
XM_017026727.1 5271 Intron XP_016882216.1
Gene
ZNF333
Gene Name
zinc finger protein 333
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300912.1 5271 Intron NP_001287841.1
NM_032433.2 5271 Intron NP_115809.1
XM_005260098.3 5271 Intron XP_005260155.1
XM_006722924.2 5271 Intron XP_006722987.1
XM_011528362.2 5271 Intron XP_011526664.1
XM_011528363.2 5271 Intron XP_011526665.1
XM_011528365.2 5271 Intron XP_011526667.1
XM_011528366.2 5271 Intron XP_011526668.1
XM_011528367.2 5271 Intron XP_011526669.1
XM_011528368.2 5271 Intron XP_011526670.1
XM_011528369.2 5271 Intron XP_011526671.1
XM_011528371.2 5271 Intron XP_011526673.1
XM_011528372.2 5271 Intron XP_011526674.1
XM_017027367.1 5271 Intron XP_016882856.1
XM_017027368.1 5271 Intron XP_016882857.1
XM_017027369.1 5271 Intron XP_016882858.1
XM_017027370.1 5271 Intron XP_016882859.1
XM_017027371.1 5271 Intron XP_016882860.1

View Full Product Details