Product Details

SNP ID
rs2253849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10451071 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTGTTCAGTTGCTAAAATGGAG[C/T]TTTTATTGCCTTTAGGTTTTCGTTG
Phenotype
MIM: 161555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRC1 PubMed Links

Gene Details

Gene
KLRC1
Gene Name
killer cell lectin like receptor C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304448.1 543 Missense Mutation AAC,AGC N29S NP_001291377.1
NM_002259.4 543 Missense Mutation AAC,AGC N29S NP_002250.1
NM_007328.3 543 Missense Mutation AAC,AGC N29S NP_015567.1
NM_213657.2 543 Missense Mutation AAC,AGC N29S NP_998822.1
NM_213658.2 543 Missense Mutation AAC,AGC N29S NP_998823.1

View Full Product Details