Product Details
- SNP ID
-
rs2734537
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:62818813 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGGCAGGCCGGGACCGGGGCTG[A/G]TACAGCCCTTCAGGTCCCCCAGGCC
- Phenotype
-
MIM: 120270
MIM: 606459
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COL9A3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116110757,rs139504978] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COL9A3
- Gene Name
- collagen type IX alpha 3
- Gene
- OGFR
- Gene Name
- opioid growth factor receptor
There are no transcripts associated with this gene.
View Full Product Details