Product Details

SNP ID
rs2734659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134444905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAAAGTCACAGGCAAATTCTTC[A/C]AGAGTTTGATCCAGGGTCACCTGGG
Phenotype
MIM: 103880
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKR1B1 PubMed Links

Gene Details

Gene
AKR1B1
Gene Name
aldo-keto reductase family 1 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001628.2 Intron NP_001619.1
XM_005250234.3 Intron XP_005250291.1

View Full Product Details