Product Details
- SNP ID
-
rs2734661
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:134444234 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACAGAGATGTCTGAAAGGGCACTG[C/G]CCTGGGGCTGCCTGTGACATGCTCT
- Phenotype
-
MIM: 103880
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AKR1B1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78265539] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKR1B1
- Gene Name
- aldo-keto reductase family 1 member B
View Full Product Details