Product Details

SNP ID
rs2753348
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:86469680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGATTTCTTGCTACCTCTTATG[C/T]AGCAGAAGGGAAAGGAAGAAGACTT
Phenotype
MIM: 603906
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCA1 PubMed Links

Gene Details

Gene
CLCA1
Gene Name
chloride channel accessory 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285.3 Intron NP_001276.2

View Full Product Details