Product Details

SNP ID
rs2854736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44375569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGGGTCCCGGGGAGGCCGGGCC[A/C]GAGACCAGAGAGCCTGCTCACTACG
Phenotype
MIM: 607759
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITGA2B PubMed Links

Gene Details

Gene
ITGA2B
Gene Name
integrin subunit alpha 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000419.4 Intron NP_000410.2
XM_011524749.1 Intron XP_011523051.1
XM_011524750.1 Intron XP_011523052.1

View Full Product Details