Product Details

SNP ID
rs2834402
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:34187610 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACATCCAGGACCTACCATCTTGAAT[G/T]TAAGTTTTTGCCCCTTGGCCCAGAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00310 PubMed Links

Gene Details

Gene
LINC00310
Gene Name
long intergenic non-protein coding RNA 310
There are no transcripts associated with this gene.

View Full Product Details