Product Details

SNP ID
rs2834608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:34728196 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCGATGAAATCAGGCTGATAGGG[A/G]ATAATGTCCCAAAACAGTCAACTGA
Phenotype
MIM: 615321
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIC6 PubMed Links

Gene Details

Gene
CLIC6
Gene Name
chloride intracellular channel 6
There are no transcripts associated with this gene.

Gene
LINC00160
Gene Name
long intergenic non-protein coding RNA 160
There are no transcripts associated with this gene.

View Full Product Details