Product Details

SNP ID
rs2856096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:1075665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCCCTCAGAGGGCAGAACCCGA[A/T]ACCACGGCCACAACGTCTGCAGCAC
Phenotype
MIM: 158370
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MUC2 PubMed Links

Gene Details

Gene
MUC2
Gene Name
mucin 2, oligomeric mucus/gel-forming
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002457.3 Intron NP_002448.3

View Full Product Details