Product Details

SNP ID
rs2144687
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:1921955 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTCACCCACACTGCAGGCTGCA[C/T]GTGTTGGCCTACAAAAAATTTATTG
Phenotype
MIM: 610171
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALML6 PubMed Links

Gene Details

Gene
CALML6
Gene Name
calmodulin like 6
There are no transcripts associated with this gene.

Gene
CFAP74
Gene Name
cilia and flagella associated protein 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304360.1 5408 UTR 3 NP_001291289.1
XM_011542333.2 5408 Intron XP_011540635.2
XM_011542335.1 5408 Intron XP_011540637.1
XM_011542336.2 5408 Intron XP_011540638.1
XM_017002641.1 5408 Intron XP_016858130.1
XM_017002642.1 5408 Intron XP_016858131.1
XM_017002643.1 5408 Intron XP_016858132.1
XM_017002644.1 5408 Intron XP_016858133.1
Gene
TMEM52
Gene Name
transmembrane protein 52
There are no transcripts associated with this gene.

View Full Product Details