Product Details

SNP ID
rs2131754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84823739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTTTGATTGTCTTGAGTGCTGC[A/G]TCAGCTAAGACAAAAACCACAGAGT
Phenotype
MIM: 612434
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRISPLD2 PubMed Links

Gene Details

Gene
CRISPLD2
Gene Name
cysteine rich secretory protein LCCL domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031476.3 Intron NP_113664.1
XM_005256190.1 Intron XP_005256247.1

View Full Product Details