Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001321038.1 | Intron | NP_001307967.1 | |||
NM_001321039.1 | Intron | NP_001307968.1 | |||
NM_013296.4 | Intron | NP_037428.3 | |||
XM_006710589.2 | Intron | XP_006710652.1 | |||
XM_011541301.2 | Intron | XP_011539603.1 | |||
XM_011541302.2 | Intron | XP_011539604.1 | |||
XM_011541303.2 | Intron | XP_011539605.1 | |||
XM_017001097.1 | Intron | XP_016856586.1 | |||
XM_017001098.1 | Intron | XP_016856587.1 |