Product Details
- SNP ID
-
rs2857277
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:178527878 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAACCCAAACAATTTGCTGTTGA[A/G]GCATAAGCAAATACATATATACATA
- Phenotype
-
MIM: 188840
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTN
PubMed Links
Gene Details
- Gene
- TTN
- Gene Name
- titin
- Gene
- TTN-AS1
- Gene Name
- TTN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details