Product Details
- SNP ID
-
rs2132101
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:8476499 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTCACTGGTACAGAGCAGATCCA[G/T]TGTGGTGAGGAGGACGGTGCTGAGG
- Phenotype
-
MIM: 160776
MIM: 607538
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MYH10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs372221867] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MYH10
- Gene Name
- myosin, heavy chain 10, non-muscle
- Gene
- NDEL1
- Gene Name
- nudE neurodevelopment protein 1 like 1
There are no transcripts associated with this gene.
View Full Product Details