Product Details

SNP ID
rs2172441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:17442222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCAGAATGTGGACAAAAATAG[C/T]AAAAATCCAGCTGGGAGTCAGTGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01549 PubMed Links

Gene Details

Gene
LINC01549
Gene Name
long intergenic non-protein coding RNA 1549
There are no transcripts associated with this gene.

View Full Product Details