Product Details
- SNP ID
-
rs2892242
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:5873797 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTGATCCTTTCCCTCTTGTCTTG[G/T]GTATTTTCTCGTGTAAGTTTTTTTG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD16
PubMed Links
Gene Details
- Gene
- ANKRD16
- Gene Name
- ankyrin repeat domain 16
View Full Product Details