Product Details

SNP ID
rs2873950
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:119828215 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTTTAGAAATACGTACAACTCTACA[A/C]CTAACCATTATCTCCTCCCCCCTTT
Phenotype
MIM: 605004 MIM: 603065
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GSK3B PubMed Links

Gene Details

Gene
GSK3B
Gene Name
glycogen synthase kinase 3 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146156.1 Intron NP_001139628.1
NM_002093.3 Intron NP_002084.2
XM_006713610.2 Intron XP_006713673.1
XM_006713611.2 Intron XP_006713674.1
Gene
NR1I2
Gene Name
nuclear receptor subfamily 1 group I member 2
There are no transcripts associated with this gene.

View Full Product Details