Product Details

SNP ID
rs2894325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33115107 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCCACCATATACTCATCACTTAT[A/G]TCTGCTTTGCAGGTAAAATTCCCTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLA-DPB2 PubMed Links

Gene Details

Gene
HLA-DPB2
Gene Name
major histocompatibility complex, class II, DP beta 2 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details