Product Details

SNP ID
rs2182661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124014451 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAAAAAGCTTGAGTTGGGATCC[G/T]TGCTCCCCTCTCTCTTTGAAGTTTC
Phenotype
MIM: 603759
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LHX2 PubMed Links

Gene Details

Gene
LHX2
Gene Name
LIM homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004789.3 Intron NP_004780.3
XM_006717323.2 Intron XP_006717386.1
Gene
LOC100505588
Gene Name
uncharacterized LOC100505588
There are no transcripts associated with this gene.

View Full Product Details