Product Details

SNP ID
rs2106806
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15629578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAGAATATGGTGGAAGAAAGTG[A/G]AGAAAAGAGAAAGCTCTTTTTTTAT
Phenotype
MIM: 300631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GS1-594A7.3 PubMed Links

Gene Details

Gene
GS1-594A7.3
Gene Name
uncharacterized LOC104798195
There are no transcripts associated with this gene.

Gene
TMEM27
Gene Name
transmembrane protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020665.5 Intron NP_065716.1
XM_017029680.1 Intron XP_016885169.1
XM_017029681.1 Intron XP_016885170.1

View Full Product Details