Product Details

SNP ID
rs2849309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:64081327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATCTTCTTGCTCATTTCAATCT[A/C]AATCAAATCTATATTAAAATATTAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00305 PubMed Links

Gene Details

Gene
LINC00305
Gene Name
long intergenic non-protein coding RNA 305
There are no transcripts associated with this gene.

View Full Product Details