Product Details

SNP ID
rs2077095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:81233620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAGGAGCAGAACAAATCAGCAC[C/G]CTGCCTGCAGGGCCTCTTTTTGGTA
Phenotype
MIM: 123295 MIM: 606614
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CKMT2 PubMed Links

Gene Details

Gene
CKMT2
Gene Name
creatine kinase, mitochondrial 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099735.1 Intron NP_001093205.1
NM_001099736.1 Intron NP_001093206.1
NM_001825.2 Intron NP_001816.2
Gene
CKMT2-AS1
Gene Name
CKMT2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
RASGRF2
Gene Name
Ras protein specific guanine nucleotide releasing factor 2
There are no transcripts associated with this gene.

View Full Product Details