Product Details

SNP ID
rs2789314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:19046343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATATGGAAACAACCTAAGTGTCCA[C/T]TGAGACATGAATGGATAAAGAAATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MALRD1 PubMed Links

Gene Details

Gene
MALRD1
Gene Name
MAM and LDL receptor class A domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142308.2 Intron NP_001135780.2
XM_011519453.2 Intron XP_011517755.1
XM_011519454.1 Intron XP_011517756.1
XM_011519455.2 Intron XP_011517757.1
XM_017016182.1 Intron XP_016871671.1
XM_017016183.1 Intron XP_016871672.1
XM_017016184.1 Intron XP_016871673.1
XM_017016185.1 Intron XP_016871674.1
XM_017016186.1 Intron XP_016871675.1

View Full Product Details