Product Details

SNP ID
rs2789790
Assay Type
Validated
NCBI dbSNP Submissions
9
Location
Chr.1:220151547 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATTTTCTTCATAACTGTTATTAA[C/G]CAGGCACTCAAGAAAACATCCAATG
Phenotype
MIM: 612801 MIM: 609275
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IARS2 PubMed Links

Gene Details

Gene
IARS2
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
RAB3GAP2
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012414.3 Intron NP_036546.2

View Full Product Details