Product Details

SNP ID
rs2082609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:224766704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGACTTCACTATGCTGTTATT[A/C]TTACTGAATGTTGAAACAGACTCCA
Phenotype
MIM: 611518
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DOCK10 PubMed Links

Gene Details

Gene
DOCK10
Gene Name
dedicator of cytokinesis 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290263.1 Intron NP_001277192.1
NM_014689.2 Intron NP_055504.2
XM_005246673.4 Intron XP_005246730.1
XM_006712617.3 Intron XP_006712680.1
XM_017004441.1 Intron XP_016859930.1
XM_017004442.1 Intron XP_016859931.1
XM_017004443.1 Intron XP_016859932.1
XM_017004444.1 Intron XP_016859933.1
XM_017004445.1 Intron XP_016859934.1
XM_017004446.1 Intron XP_016859935.1
XM_017004447.1 Intron XP_016859936.1

View Full Product Details